Aquatic Block™ Serum-Free PBS, Azide-Free (with Surfactant)
Supplier: ARLINGTON SCIENTIFIC INC.
A non-mammalian blocking reagent supplied in PBS for diagnostic systems involving membrane or other solid phase material.
- Imparts superior stability and flow characteristics
- Utilizes proteins that do not interact with mammalian antibodies
- Supplied in HDPE plastic
Azide-free, with surfactant.
Applications: Lateral flow, ELISA.