Aqua Block™ PBS
Supplier: ARLINGTON SCIENTIFIC INC.
A non-mammalian blocking reagent supplied in PBS for diagnostic systems involving membrane or other solid phase material.
- Optimized for superior performance in super sensitive assay systems
- Utilizes proteins that do not interact with mammalian antibodies
- Supplied in HDPE plastic
Preservative - 0.09% sodium azide.
Applications: Western blot/EIA/Chemiluminescence.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...