Orthophosphoric acid 85% w/w ACS

Supplier: AQUA SOLUTIONS MS

Synonyms: Phosphoric acid

P2380-4L P2380-500ML P2380-2.5L
101204-140EA 407.63 USD
101204-140 101204-490 101203-712
Orthophosphoric acid 85% w/w ACS
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

Purity (v/v) 25%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR