Eisco Heavy Wall Test Tubes
Supplier: Wards
CH0719A CH0719B CH0719C CH0719D CH0719E CH0719F
470354-588PK 83 USD
470354-588 470354-590 470354-592 470354-594 470354-596 470354-598
Eisco Heavy Wall Test Tubes
Tubes Test Tubes
Durable heavy wall test tubes.
  • Borosilicate glass 3.3, heavy wall, with rim
  • Strong resistance to heat and chemicals
  • 1.5 mm wall thickness
These products have a heavy wall with a rim. Has strong resistance to heat and chemicals. Comes in bulk packs. Comes in multiple different capacities.