Myrcene, natural isolate
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
7-Methyl-3-methyleneocta-1,6-diene
RCCT9085-20C6
RCCT9085-750C6
RCCT9085-100C6
RCCT9085-100C6EA
124.84
USD
RCCT9085-100C6
RCCT9085-20C6
RCCT9085-750C6
Myrcene, natural isolate
Myrcene
Formula:
C₁₀H₁₆ MW: 136.24 g/mol |
MDL Number:
MFCD00008908 CAS Number: 123-35-3 UN: 2319 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...