β-Pinene, natural isolate

Supplier: Ricca Chemical
Danger

Synonyms: Nopinenebeta-PinenePseudopinene2(10)-PineneTerebenthene

RCCT9029-100C6 RCCT9029-750C6 RCCT9029-20C6
RCCT9029-20C6EA 21.4 USD
RCCT9029-20C6 RCCT9029-100C6 RCCT9029-750C6
β-Pinene, natural isolate
β-Pinene
Formula: C₁₀H₁₆
MW: 136.23 g/mol
MDL Number: MFCD00064767
CAS Number: 127-91-3
UN: 2319
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR