Farnesene, natural isolate

Supplier: Ricca Chemical

Synonyms: Farnesene, mixture of isomers(3E,6E)-3,7,11-trimethyldodeca-1,3,6,10-tetraene

RCCT9014-100C6 RCCT9014-20C6 RCCT9014-750C6
RCCT9014-750C6EA 6509.31 USD
RCCT9014-750C6 RCCT9014-20C6 RCCT9014-100C6
Farnesene, natural isolate
Farnesene
Formula: C₁₅H₂₄
MW: 204.35 g/mol
MDL Number: MFCD00036650
CAS Number: 502-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR