(±)-Linalool, natural isolate

Supplier: Ricca Chemical
Warning

Synonyms: 3,7-Dimethylocta-1,6-dien-3-ol(±)-3,7-Dimethyl-1,6-octadien-3-ol

RCCT9009-100C6 RCCT9009-750C6 RCCT9009-20C6
RCCT9009-100C6EA 89.17 USD
RCCT9009-100C6 RCCT9009-750C6 RCCT9009-20C6
(±)-Linalool, natural isolate
(±)-Linalool
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00008906
CAS Number: 78-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR