Phosphonate Test Kit
Supplier: LA MOTTE CHEMICAL PRODUCTS CO
7625-DR-01
66170-470EA 131.99 USD
66170-470
Phosphonate Test Kit
Phosphonate Test Kit
This kit tests for Phosphonate using the Direct Reading Titrator. Range and Sensitivity is 0-20 ppm/0.4 ppm NaAMP.