3,5-Difluoro-2-methoxyphenylboronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(3,5-Difluoro-2-methoxyphenyl)boronic acid, 3,5-Difluoro-2-methoxybenzeneboronic acid
11691-5G
11691-25G
77359-720EA
131.84
USD
77359-720
77371-700
3,5-Difluoro-2-methoxyphenylboronic acid ≥95%
3,5-Difluoro-2-methoxyphenylboronic acid
Caution: for Research Use Only
Formula:
C₇H₇BF₂O₃ MW: 187.94 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03095353 CAS Number: 737000-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...