VACUUBRAND® RZ 2.5 Rotary Vane Pump
Supplier: BrandTech
VACUUBRAND®
20798126
77322-838EA 3054.46 USD
77322-838
VACUUBRAND® RZ 2.5 Rotary Vane Pump
Pumps Vacuum Rotary Vane Pumps
The RZ 2.5 from VACUUBRAND delivers impressive performances in a compact design.
  • Powerful ultimate vacuum even with gas ballast open
  • High-performing, energy efficient motor
  • Whisper quiet with minimal vibrations
  • Small footprint and easy maintenance
The two-stage RZ 2.5 rotary vacuum pump is the optimal choice for any laboratory process demanding low ultimate vacuum at a medium gas flow rate.
The compact, telescopic design makes required maintenance effortless, and the large oil volume capacity provides extended intervals between oil changes.
Common applications include lyophilization, fine vacuum distillation, and evacuation of small volumes.
Certifications: CE, cTUVus, UKCA, WEEE, China RoHS 40.
Delivery information: Includes ready-to-use oil-filled pump, operations manual, and US plug.