4-(5-Bromo-2-fluorophenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

92450-500MG 92450-1G 92450-5G 92450-25G
77335-148EA 223.51 USD
77335-148 77359-240 77371-188 77391-664
4-(5-Bromo-2-fluorophenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(5-Bromo-2-fluorophenyl)tetrahydro-2H-pyran-4-ol

Caution: for Research Use Only

Formula: C₁₁H₁₂BrFO₂
Storage Temperature: Ambient
CAS Number: 1262037-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR