Ethyl-4-chloro-3-fluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(4-chloro-3-fluorophenyl)-2-oxoacetate

82448-500MG 82448-1G 82448-5G 82448-25G
77334-670EA 151.41 USD
77334-670 77345-432 77370-692 77391-336
Ethyl-4-chloro-3-fluorobenzoylformate ≥95%
Ethyl 4-chloro-3-fluorobenzoylformate

Caution: for Research Use Only

Formula: C₁₀H₈ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD06201712
CAS Number: 845790-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR