2,3-Dimethyl-4-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

74304-500MG 74304-1G 74304-5G
77333-992EA 342.99 USD
77333-992 77358-174 77382-070
2,3-Dimethyl-4-(methylthio)phenol ≥97%
2,3-Dimethyl-4-(methylthio)phenol

Caution: for Research Use Only

Formula: C₉H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD24714166
CAS Number: 7379-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR