5-Fluoro-2-methyl-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

72798-500MG 72798-1G 72798-5G
77335-914EA 367.71 USD
77335-914 77357-990 77381-908
5-Fluoro-2-methyl-3-(methylthio)benzoic acid ≥97%
5-Fluoro-2-methyl-3-(methylthio)benzoic acid

Caution: for Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD34764564
CAS Number: 2807454-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR