2,2,2-Trifluoro-1-(4-isopropoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

72620-500MG 72620-1G 72620-5G 72620-25G
77333-772EA 120.51 USD
77333-772 77343-816 77369-788 77381-850
2,2,2-Trifluoro-1-(4-isopropoxyphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-isopropoxyphenyl)ethanone

Caution: for Research Use Only

Formula: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD17260509
CAS Number: 1339633-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR