1-Bromo-3-(methylsulfonyl)-5-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

66452-500MG 66452-1G 66452-5G 66452-25G
77332-796EA 107.12 USD
77332-796 77343-154 77368-844 77380-976
1-Bromo-3-(methylsulfonyl)-5-(trifluoromethoxy)benzene ≥95%
1-Bromo-3-(methylsulfonyl)-5-(trifluoromethoxy)benzene

Caution: for Research Use Only

Formula: C₈H₆BrF₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD32701834
CAS Number: 2749436-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR