4-Methyl-3-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66039-500MG 66039-1G 66039-5G
77335-608EA 360.5 USD
77335-608 77356-968 77380-906
4-Methyl-3-nitro-1,1'-biphenyl ≥97%
4-Methyl-3-nitro-1,1'-biphenyl

Caution: for Research Use Only

Formula: C₁₃H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00222364
CAS Number: 80726-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR