2,2,2,2',4'-Pentafluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2,4-Difluorophenyl)-2,2,2-trifluoroethanone

63836-500MG 63836-1G 63836-5G 63836-25G
77332-008EA 92.7 USD
77332-008 77342-472 77368-102 77380-378
2,2,2,2',4'-Pentafluoroacetophenone ≥95%
2,2,2,2',4'-Pentafluoroacetophenone

Caution: for Research Use Only

Formula: C₈H₃F₅O
Storage Temperature: Ambient
MDL Number: MFCD02260842
CAS Number: 886371-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR