1-(5-Ethyl-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63185-500MG 63185-1G 63185-5G
77331-846EA 345.05 USD
77331-846 77356-138 77380-180
1-(5-Ethyl-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(5-Ethyl-2-fluorophenyl)-2,2,2-trifluoroethanone

Caution: for Research Use Only

Formula: C₁₀H₈F₄O
Storage Temperature: Ambient
MDL Number: MFCD28061684
CAS Number: 1823911-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR