4-Ethoxy-2-fluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62649-500MG 62649-1G 62649-5G
77339-186EA 560.32 USD
77339-186 77355-938 77379-996
4-Ethoxy-2-fluoro-5-methylbenzaldehyde ≥97%
4-Ethoxy-2-fluoro-5-methylbenzaldehyde

Caution: for Research Use Only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD34760957
CAS Number: 2092769-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR