4-Chloro-2-fluoro-3-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

61971-500MG 61971-1G 61971-5G
77335-484EA 355.35 USD
77335-484 77355-726 77379-772
4-Chloro-2-fluoro-3-(methylthio)phenol ≥97%
4-Chloro-2-fluoro-3-(methylthio)phenol

Caution: for Research Use Only

Formula: C₇H₆ClFOS
Storage Temperature: Ambient
MDL Number: MFCD34760658
CAS Number: 2707341-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR