1-(2,3-Difluoro-4-methoxyphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

61748-500MG 61748-1G 61748-5G 61748-25G
77331-380EA 256.47 USD
77331-380 77355-652 77367-522 77389-018
1-(2,3-Difluoro-4-methoxyphenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,3-Difluoro-4-methoxyphenyl)-2,2,2-trifluoroethanone

Caution: for Research Use Only

Formula: C₉H₅F₅O₂
Storage Temperature: Ambient
MDL Number: MFCD20528783
CAS Number: 1356109-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR