3-(Benzyloxy)-5-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61414-500MG 61414-1G 61414-5G
77336-046EA 374.92 USD
77336-046 77355-482 77379-540
3-(Benzyloxy)-5-isopropylbenzoic acid ≥97%
3-(Benzyloxy)-5-isopropylbenzoic acid

Caution: for Research Use Only

Formula: C₁₇H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD34760332
CAS Number: 2764733-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR