2,2,2-Trifluoro-1-(2-fluoro-6-methoxy-3-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

61097-500MG 61097-1G 61097-5G 61097-25G
77331-130EA 208.06 USD
77331-130 77355-348 77367-324 77388-838
2,2,2-Trifluoro-1-(2-fluoro-6-methoxy-3-methylphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(2-fluoro-6-methoxy-3-methylphenyl)ethanone

Caution: for Research Use Only

Formula: C₁₀H₈F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34760136
CAS Number: 2755722-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR