2-Ethoxy-3-fluoro-6-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60850-500MG 60850-1G 60850-5G
77338-020EA 459.38 USD
77338-020 77355-286 77379-356
2-Ethoxy-3-fluoro-6-methoxybenzaldehyde ≥97%
2-Ethoxy-3-fluoro-6-methoxybenzaldehyde

Caution: for Research Use Only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD34759987
CAS Number: 2386333-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR