1,2,5-Tribromo-3-methylbenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
60613-500MG
60613-1G
60613-5G
60613-25G
77330-954EA
120.51
USD
77330-954
77343-770
77367-154
77379-260
1,2,5-Tribromo-3-methylbenzene ≥97%
1,2,5-Tribromo-3-methylbenzene
Caution: for Research Use Only
Formula:
C₇H₅Br₃ MW: 328.827 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18400101 CAS Number: 140141-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...