4-Chloro-1,3,5-trimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

60538-500MG 60538-1G 60538-5G 60538-25G
77330-926EA 181.28 USD
77330-926 77346-866 77367-128 77388-686
4-Chloro-1,3,5-trimethyl-1H-pyrazole ≥97%
4-Chloro-1,3,5-trimethyl-1H-pyrazole

Caution: for Research Use Only

Formula: C₆H₉ClN₂
Storage Temperature: Ambient
MDL Number: MFCD00464340
CAS Number: 56079-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR