2-Fluoro-6-isopropoxy-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60514-500MG 60514-1G 60514-5G 60514-25G
77330-908EA 90.64 USD
77330-908 77342-434 77367-112 77379-218
2-Fluoro-6-isopropoxy-3-methoxybenzaldehyde ≥97%
2-Fluoro-6-isopropoxy-3-methoxybenzaldehyde

Caution: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759793
CAS Number: 2918828-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR