3-Bromo-N,5-dimethoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

60483-500MG 60483-1G 60483-5G
77330-896EA 319.3 USD
77330-896 77355-112 77379-194
3-Bromo-N,5-dimethoxy-N-methylbenzamide ≥97%
3-Bromo-N,5-dimethoxy-N-methylbenzamide

Caution: for Research Use Only

Formula: C₁₀H₁₂BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD34759772
CAS Number: 774239-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR