1-Chloro-3-ethyl-2-fluoro-4-iodobenzene ≥95%
Supplier: AOBChem USA
|
Warning
|
60466-500MG
60466-1G
60466-5G
77339-554EA
596.37
USD
77339-554
77355-108
77379-188
1-Chloro-3-ethyl-2-fluoro-4-iodobenzene ≥95%
1-Chloro-3-ethyl-2-fluoro-4-iodobenzene
Caution: for Research Use Only
Formula:
C₈H₇ClFI MW: 284.497 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759759 CAS Number: 2708074-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...