4-Fluoro-2-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

60256-500MG 60256-1G 60256-5G 60256-25G
77330-824EA 208.06 USD
77330-824 77355-022 77367-034 77388-620
4-Fluoro-2-(methylthio)benzaldehyde ≥95%
4-Fluoro-2-(methylthio)benzaldehyde

Caution: for Research Use Only

Formula: C₈H₇FOS
Storage Temperature: Ambient
MDL Number: MFCD12022603
CAS Number: 183951-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR