3-Ethoxy-4-(trifluoromethoxy)benzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
59911-500MG
59911-1G
59911-25G
59911-5G
77330-744EA
287.37
USD
77330-744
77354-906
77388-560
77379-010
3-Ethoxy-4-(trifluoromethoxy)benzoic acid ≥97%
3-Ethoxy-4-(trifluoromethoxy)benzoic acid
Caution: for Research Use Only
Formula:
C₁₀H₉F₃O₄ Storage Temperature: Ambient |
MDL Number:
MFCD34759480 CAS Number: 2764732-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...