3-Chloro-4-ethoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

59135-500MG 59135-1G 59135-5G
77336-250EA 384.19 USD
77336-250 77354-640 77378-728
3-Chloro-4-ethoxy-5-methylbenzaldehyde ≥97%
3-Chloro-4-ethoxy-5-methylbenzaldehyde

Caution: for Research Use Only

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD06246190
CAS Number: 883522-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR