4-Hydroxy-2,5-dimethylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
58454-500MG
58454-1G
58454-5G
58454-25G
77330-290EA
167.89
USD
77330-290
77346-286
77366-536
77388-260
4-Hydroxy-2,5-dimethylbenzoic acid ≥97%
4-Hydroxy-2,5-dimethylbenzoic acid
Caution: for Research Use Only
Formula:
C₉H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD09999327 CAS Number: 27021-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...