3-hydroxy-3-(p-tolyl)propanoic acid ≥97%

Supplier: AOBChem USA

Warning

58199-500MG 58199-1G 58199-5G 58199-25G
77330-172EA 229.69 USD
77330-172 77354-200 77366-446 77388-164
3-hydroxy-3-(p-tolyl)propanoic acid ≥97%
3-hydroxy-3-(p-tolyl)propanoic acid

Caution: for Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD16302027
CAS Number: 61752-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR