4-Methyl-[1,1'-biphenyl]-2-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58103-500MG 58103-1G 58103-5G 58103-25G
77330-130EA 268.83 USD
77330-130 77354-146 77366-414 77388-136
4-Methyl-[1,1'-biphenyl]-2-carbaldehyde ≥97%
4-Methyl-[1,1'-biphenyl]-2-carbaldehyde

Caution: for Research Use Only

Formula: C₁₄H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD24674578
CAS Number: 2928-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR