Methyl 2-(benzyloxy)-4-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

57776-500MG 57776-1G 57776-5G
77340-036EA 722.03 USD
77340-036 77354-022 77378-228
Methyl 2-(benzyloxy)-4-fluoro-5-methylbenzoate ≥97%
Methyl 2-(benzyloxy)-4-fluoro-5-methylbenzoate

Caution: for Research Use Only

Formula: C₁₆H₁₅FO₃
Storage Temperature: Ambient
MDL Number: MFCD34758294
CAS Number: 2755717-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR