3-(3-Chloro-5-(trifluoromethyl)phenyl)-3-hydroxypropanoic acid ≥97%

Supplier: AOBChem USA

Warning

55934-500MG 55934-1G 55934-5G
77339-074EA 548.99 USD
77339-074 77353-488 77377-736
3-(3-Chloro-5-(trifluoromethyl)phenyl)-3-hydroxypropanoic acid ≥97%
3-(3-Chloro-5-(trifluoromethyl)phenyl)-3-hydroxypropanoic acid

Caution: for Research Use Only

Formula: C₁₀H₈ClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34757649
CAS Number: 2229091-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR