Ethyl 2-ethoxy-4-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

54638-500MG 54638-1G 54638-5G
77335-752EA 364.62 USD
77335-752 77353-156 77377-400
Ethyl 2-ethoxy-4-(trifluoromethyl)benzoate ≥95%
Ethyl 2-ethoxy-4-(trifluoromethyl)benzoate

Caution: for Research Use Only

Formula: C₁₂H₁₃F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34535491
CAS Number: 939983-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR