1-Bromo-2,5-dichloro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

54141-500MG 54141-1G 54141-5G 54141-25G
77329-192EA 204.97 USD
77329-192 77353-008 77365-480 77387-468
1-Bromo-2,5-dichloro-3-iodobenzene ≥97%
1-Bromo-2,5-dichloro-3-iodobenzene

Caution: for Research Use Only

Formula: C₆H₂BrCl₂I
Storage Temperature: Ambient
MDL Number: MFCD32659897
CAS Number: 1936731-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR