1-(2-Bromo-3-ethoxy-6-fluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

52883-500MG 52883-1G 52883-5G 52883-25G
77328-826EA 145.23 USD
77328-826 77345-078 77365-150 77387-230
1-(2-Bromo-3-ethoxy-6-fluorophenyl)ethanol ≥97%
1-(2-Bromo-3-ethoxy-6-fluorophenyl)ethanol

Caution: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34178639
CAS Number: 2643367-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR