3-(Benzyloxy)-4-fluoro-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

52511-500MG 52511-1G 52511-25G 52511-5G
77328-716EA 298.7 USD
77328-716 77352-402 77387-146 77376-682
3-(Benzyloxy)-4-fluoro-5-(methylthio)benzoic acid ≥97%
3-(Benzyloxy)-4-fluoro-5-(methylthio)benzoic acid

Caution: for Research Use Only

Formula: C₁₅H₁₃FO₃S
Storage Temperature: Ambient
MDL Number: MFCD33404072
CAS Number: 2635937-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR