Ethyl 4-bromo-1-fluoro-2-naphthoate ≥97%

Supplier: AOBChem USA

Warning

52344-500MG 52344-1G 52344-5G 52344-25G
77328-654EA 168.92 USD
77328-654 77346-328 77364-998 77387-100
Ethyl 4-bromo-1-fluoro-2-naphthoate ≥97%
Ethyl 4-bromo-1-fluoro-2-naphthoate

Caution: for Research Use Only

Formula: C₁₃H₁₀BrFO₂
MW: 297.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530171
CAS Number: 2734778-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR