4-Bromo-1-fluoro-2-iodonaphthalene ≥97%

Supplier: AOBChem USA

Warning

52341-500MG 52341-1G 52341-5G 52341-25G
77328-652EA 98.88 USD
77328-652 77342-710 77364-996 77376-614
4-Bromo-1-fluoro-2-iodonaphthalene ≥97%
4-Bromo-1-fluoro-2-iodonaphthalene

Caution: for Research Use Only

Formula: C₁₀H₅BrFI
MW: 350.954 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404047
CAS Number: 2103228-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR