2,6-Dichloro-3-formylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
52073-500MG
52073-1G
52073-5G
52073-25G
77328-612EA
136.99
USD
77328-612
77344-536
77364-946
77387-072
2,6-Dichloro-3-formylbenzoic acid ≥97%
2,6-Dichloro-3-formylbenzoic acid
Caution: for Research Use Only
Formula:
C₈H₄Cl₂O₃ MW: 219.022 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24488418 CAS Number: 189046-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...