6-Methyl-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

51888-500MG 51888-1G 51888-5G 51888-25G
77328-570EA 211.15 USD
77328-570 77352-170 77364-908 77387-048
6-Methyl-[1,1'-biphenyl]-3-carboxylic acid ≥97%
6-Methyl-[1,1'-biphenyl]-3-carboxylic acid

Caution: for Research Use Only

Formula: C₁₄H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11577105
CAS Number: 401573-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR