Ethyl 2-ethoxy-4-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

48386-500MG 48386-1G 48386-5G 48386-25G
77327-560EA 261.62 USD
77327-560 77351-152 77363-902 77386-336
Ethyl 2-ethoxy-4-fluorobenzoate ≥97%
Ethyl 2-ethoxy-4-fluorobenzoate

Caution: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD28680209
CAS Number: 939983-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR