(3-Chloro-2,4-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

47709-500MG 47709-1G 47709-5G 47709-25G
77327-402EA 95.79 USD
77327-402 77342-612 77363-750 77375-308
(3-Chloro-2,4-difluorophenyl)(methyl)sulfane ≥97%
(3-Chloro-2,4-difluorophenyl)(methyl)sulfane

Caution: for Research Use Only

Formula: C₇H₅ClF₂S
Storage Temperature: Ambient
MDL Number: MFCD28751305
CAS Number: 1807122-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR