(3-Chloro-2-fluoro-5-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

47272-500MG 47272-1G 47272-5G 47272-25G
77327-316EA 128.75 USD
77327-316 77344-246 77363-656 77375-220
(3-Chloro-2-fluoro-5-iodophenyl)methanol ≥97%
(3-Chloro-2-fluoro-5-iodophenyl)methanol

Caution: for Research Use Only

Formula: C₇H₅ClFIO
Storage Temperature: Ambient
MDL Number: MFCD34178524
CAS Number: 1000162-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR